Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LY 456236 hydrochloride
GREENER_CHOICESDP

Catalog No. 239050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-905-0 50 mg
23-901-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-905-0 Supplier R&D Systems™ Supplier No. 2390/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective mGlu1 antagonist

Selective mGlu1 receptor antagonist (IC50 values are 143 nM and > 10 μM for mGlu1 and mGlu5 receptors respectively). Reduces hyperalgesic behavior induced by formalin in both mouse and rat with ED50 values of 28 and 16.3 mg/kg respectively.

Chemical Identifiers

CAS 338736-46-2
Molecular Formula C16H16ClN3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 317.77
Purity >99%
Molecular Weight (g/mol) 317.773
Chemical Name or Material LY 456236 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.