Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49A/D Monoclonal Antibody (eBio12A8 (12A8)), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010858
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-108-58 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-108-58 Supplier Invitrogen™ Supplier No. 12578381
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The eBio12A8 antibody reacts with Ly-49A and Ly-49D. The Ly-49 family of NK-cell receptors, members of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains which bind to MHC class I alloantigens. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. Ly-49D is expressed on subsets of natural killer cells in C57BL/6, C3H/He, and SJL mice, but not DBA/2, AKR, CBA/J, or BALB/c mice. The Ly-49D antigen has not been detected on NK-1.1+ T cells. The Ly-49D receptor has been implicated in mediation of NK cell cytolysis. Ly-49D is an activating receptor while Ly-49A is a inhibitory receptor. eBio12A8 does not react with BALB/c, DBA/2, AKR, or CBA/J Ly-D49D. Applications Reported: This eBio12A8 (12A8) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio12A8 (12A8) antibody has been tested by flow cytometric analysis of mouse spleen cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 5...

The Ly-49 family of NK-cell receptors, members of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains which bind to MHC class I alloantigens. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. Ly-49D is expressed on subsets of natural killer cells in C57BL/6, C3H/He, and SJL mice, but not DBA/2, AKR, CBA/J, or BALB/c mice. The Ly-49D antigen has not been detected on NK-1.1+ T cells. The Ly-49D receptor has been implicated in mediation of NK cell cytolysis. Ly-49D is an activating receptor while Ly-49A is a inhibitory receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49A/D
Applications Flow Cytometry
Classification Monoclonal
Clone eBio12A8 (12A8)
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra1
Gene Accession No. P20937, Q60651
Gene Alias A1; CH29-493D4.3; Chok; class I MHC receptor KLRA22; killer cell lectin-like receptor 4; killer cell lectin-like receptor subfamily A member 32; killer cell lectin-like receptor subfamily A member 33; killer cell lectin-like receptor, subfamily A, member 1; killer cell lectin-like receptor, subfamily A, member 4; Klra1; Klra22; Klra32; Klra33; Klra4; Ly49a; ly-49a; Ly49d; Ly-49d; Ly49-d; Ly49o>129>; ly49r>129>; Ly49v; lymphocyte antigen 49a; lymphocyte antigen 49d; natural killer cell receptor Ly49A; novel killer cell lectin-like receptor subfamily A protein; t lymphocyte antigen A1; T-cell surface glycoprotein Ly-49D; T-cell surface glycoprotein YE1/48
Gene Symbols Klra1, Klra4
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16627, 16635
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.