Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49F Monoclonal Antibody (HBF-719), Biotin
GREENER_CHOICE

Catalog No. PIMA528665
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528665 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528665 Supplier Invitrogen™ Supplier No. MA528665
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ly-49F Monoclonal Antibody for Flow

Ly49 receptors (Lectin like receptor) exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49F
Applications Flow Cytometry
Classification Monoclonal
Clone HBF-719
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4
Gene Klra6
Gene Accession No. Q60653
Gene Alias Killer cell lectin-like receptor 6; killer cell lectin-like receptor, subfamily A, member 6; Klra6; Ly-49; Ly49F; Ly-49f; Ly49-f; lymphocyte antigen 49f; T-cell surface glycoprotein Ly-49F
Gene Symbols Klra6
Host Species Mouse
Immunogen CHO-K1 cells transfected with B6 allele of Ly49F gene.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16637
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.