Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49F Monoclonal Antibody (HBF-719), FITC

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MLY49F01

Encompass_Preferred

Catalog No. 50-113-7528


Only null left
Add to Cart

Description

Description

Ly-49F Monoclonal Antibody for Flow

Ly49 receptors (Lectin like receptor) exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-49F
Monoclonal
0.5 mg/mL
PBS with <0.1% sodium azide; pH 7.4
Q60653
Klra6
Mouse Ly49F. from CHO-K1 cells transfected with B6 allele of Ly49F gene.
100 μg
Primary
Mouse
Antibody
IgG1 κ
Flow Cytometry
HBF-719
FITC
Klra6
Killer cell lectin-like receptor 6; killer cell lectin-like receptor, subfamily A, member 6; Klra6; Ly-49; Ly49F; Ly-49f; Ly49-f; lymphocyte antigen 49f; T-cell surface glycoprotein Ly-49F
Mouse
purified
RUO
16637
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.