Learn More
Invitrogen™ Ly-49G Monoclonal Antibody (AT-8 (AT8)), Biotin, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 13588582

Description
Description: The AT8 monoclonal antibody reacts with the mouse Ly-49G receptor from 129, FVB, BALB/c, C3H, CBA, SJL and A/J, but not C57Bl/6 mice. Ly49G, a member of the Ly49 family is expressed by NK cell subset. Applications Reported: The AT-8 (AT8) antibody has been reported for use in flow cytometric analysis. Applications Tested: The AT-8 (AT8) antibody has been tested by flow cytometric analysis of BALB/c mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Mouse Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
Specifications
Ly-49G | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q60654 | |
Klra7 | |
Affinity chromatography | |
RUO | |
16638 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
AT-8 (AT8) | |
Biotin | |
Klra7 | |
Killer cell lectin-like receptor 7; killer cell lectin-like receptor, subfamily A, member 7; Klra7; LGL-1; Ly49g; ly-49g; Ly49-g; Ly-49G.1; Ly-49G.2; Ly-49G.3; Ly49G.4; Ly49g1; Ly49g2; Ly49g4; Lymphocyte antigen 49g; T-cell surface glycoprotein Ly-49G | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.