Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49G2 Rat anti-Mouse, Brilliant Violet 421, Clone: 4D11, BD Optibuild™

Catalog No. BDB742879 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742879 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742879 Supplier BD Biosciences Supplier No. 742879
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 4D11 antibody specifically recognizes Ly-49G2 (also known as LGL-1), an inhibitory receptor which is expressed on subsets of natural killer (NK) cells and DX5-positive T lymphocytes (NK-T cells) in all strains tested (e.g., AKR/N, BALB/c, C3H/HeJ, C57BL/6, CBA/J, DBA/2, SJL, 129) and on a population of memory CD8+ T lymphocytes in C57BL/6 mice. Cross-reaction of 4D11 antibody to Ly-49A[B6], Ly-49A[BALB], and Ly-49T[129/J] inhibitory receptors and Ly-49L[CBA/J] activating receptor has been reported. The proportion of NK-T cells expressing Ly-49A and Ly-49G2 is higher (2-5 fold) in thymus than in liver (immature and mature NK-T cells, respectively), and there is evidence that down-regulation of Ly-49 receptor expression is necessary for normal NK-T-cell development to occur. Most NK cells express a single allele of Ly-49A and/or Ly-49G2, although occasionally they may express more than one allele. The Ly-49 family of NK-cell receptors, members of the C-type lectin superfamily, are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains, which bind to MHC class I alloantigens. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. Binding of Ly-49G[B6]-expressing transfectants to H-2Dd+/H-2Ld+ ConA blasts has been demonstrated, and H-2D[d]-expressing target cells inhibit the lytic activity of Ly-49G2-expressing NK cells. The levels of the Ly-49 inhibitory receptors are down-regulated by their ligands in vivo, and the various levels of expression of a Ly-49 inhibitory receptor may affect the specificity of NK cells. Ly-49G2[+] NK cells are able to lyse target tumor cells expressing H-2[a] and H-2[b] MHC class I antigens in vitro, and they mediate allogeneic and hybrid resistance to H-2[b] bone marrow transplantation. The Ly-49A[BALB] and Ly-49A[B6] alloantigens bind to MHC class I antigens of the d and k haplotypes, and Ly-49A[+] IL-2-activated NK cells are unable to lyse target cells expressing H-2[d] and H-2[k]. In vitrostudies suggest that the Ly-49G2 and Ly-49A receptors mediate negative regulation of NK-cell cytolytic activity via tyrosine phosphorylation of their ITIMs ( Immunoreceptor Tyrosine-based Inhibitory M The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49G2
Classification Monoclonal
Clone 4D11
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias LGL-1, Klra7
Host Species Rat
Immunogen Large granular lymphocytes (LGL) enriched from C57BL/6N mouse liver
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.