Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49H Mouse anti-Mouse, Brilliant Violet 510, Clone: 3D10, BD Optibuild™

Catalog No. BDB744261 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744261 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744261 Supplier BD Biosciences Supplier No. 744261
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3D10 monoclonal antibody specifically binds to mouse Lymphocyte antigen 49H (Ly-49H; also known as Klra8 or Killer cell lectin-like receptor 8). The 3D10 antibody does not crossreact with related molecules such as Ly-49A, C, D or G2. Ly-49H is a type II transmembrane protein and a member of the Ly-49 C-type lectin multigene family of receptors expressed by NK cells. Cell surface Ly-49H is expressed by a subset of NK cells but not by NKT cells. Ly-49H is expressed by C57BL/6 and NWNA but not by BALB/c or DBA/2 mouse NK cells. Cell surface Ly-49H presents as a ñ110 kDa disfulfide-linked homodimer and associates with signaling subunits such as DAP10 and DAP12 for optimal transduction of intracellular activation signals. Crosslinking of Ly-49H with the 3D10 antibody reportedly induces NK cell cytotoxicity and cytokine production. Ly-49H recognizes the mouse cytomegalovirus m157 glycoprotein that is expressed by infected cells and is required for protection against cytomegalovirus infection.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49H
Classification Monoclonal
Clone 3D10
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly49h, Lymphocyte antigen 49H, Klra8, Cmv1, Cmv-1
Host Species Mouse
Immunogen Mouse Ly-49A/H Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.