Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49H Mouse anti-Mouse, Brilliant Violet 786, Clone: 3D10, BD Optibuild™

Catalog No. BDB744265 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744265 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744265 Supplier BD Biosciences Supplier No. 744265
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3D10 monoclonal antibody specifically binds to mouse Lymphocyte antigen 49H (Ly-49H; also known as Klra8 or Killer cell lectin-like receptor 8). The 3D10 antibody does not crossreact with related molecules such as Ly-49A, C, D or G2. Ly-49H is a type II transmembrane protein and a member of the Ly-49 C-type lectin multigene family of receptors expressed by NK cells. Cell surface Ly-49H is expressed by a subset of NK cells but not by NKT cells. Ly-49H is expressed by C57BL/6 and NWNA but not by BALB/c or DBA/2 mouse NK cells. Cell surface Ly-49H presents as a ñ110 kDa disfulfide-linked homodimer and associates with signaling subunits such as DAP10 and DAP12 for optimal transduction of intracellular activation signals. Crosslinking of Ly-49H with the 3D10 antibody reportedly induces NK cell cytotoxicity and cytokine production. Ly-49H recognizes the mouse cytomegalovirus m157 glycoprotein that is expressed by infected cells and is required for protection against cytomegalovirus infection.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49H
Classification Monoclonal
Clone 3D10
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly49h, Lymphocyte antigen 49H, Klra8, Cmv1, Cmv-1
Host Species Mouse
Immunogen Mouse Ly-49A/H Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.