Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49I Monoclonal Antibody (YLI-90), FITC, eBioscience™, Invitrogen™

Catalog No. 509906
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-990-6 500 μg
50-990-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-990-6 Supplier Invitrogen™ Supplier No. 11589585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The YLI-90 monoclonal antibody reacts with the B6 mouse Ly-49I receptor. Ly49I, a member of the Ly49 family with inhibitory function, is a homodimer type II transmembrane protein and contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM). YLI-90 binds to subpopulations of NK, NKT and T cell lineages in the C57Bl/6 strain. This antibody does not bind to cells from BALB/c, SJL or AKR/J mice. Applications Reported: YLI-90 has been reported for use in flow cytometric analysis. Applications Tested: The YLI-90 antibody has been tested by flow cytometric analysis of C57Bl/6 and BALB/c mouse splenocytes and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49I
Applications Flow Cytometry
Classification Monoclonal
Clone YLI-90
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra9
Gene Accession No. 0
Gene Alias 5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor subfamily A, member 9; Klra3; Klra9; Ly-49; Ly49C; Ly-49c; Ly49I; Ly49I receptor; Ly-49I receptor; LY49I1; Lymphocyte antigen 49c; Nk2.1; T-cell surface glycoprotein Ly-49C
Gene Symbols Klra9
Host Species Mouse
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16640
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.