Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-49I Mouse anti-Mouse, BB700, Clone: YLI-90, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 746107
Description
Ly-49I Monoclonal antibody specifically detects Ly-49I in Mouse samples. It is validated for Flow Cytometry.
Specifications
Ly-49I | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
YLI-90 | |
Brilliant Blue 700 (BB700) | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction