Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-49I Mouse, anti-Mouse, R718, Clone: YLI-90, BD OptiBuild™

Catalog No. BDB752179 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752179 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752179 Supplier BD Biosciences Supplier No. 752179
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The YLI-90 monoclonal antibody specifically reacts with the B6 alloantigen of Ly-49I, an inhibitory receptor which is expressed on subsets of natural killer (NK) cells, NK-1.1+ (or DX5+) T lymphocytes (NK-T cells), and on a population of memory CD8+ T lymphocytes in C57BL/6 mice, but not AKR/J, BALB/c, DBA/1, or SJL mice. The Ly-49 family of NK-cell receptors are disulfide-linked type-II transmembrane protein homodimers with extracellular carbohydrate-recognition domains (CRD). Ly-49I is closely related to Ly-49C[B6] and Ly49C[BALB], and these three alloantigens are recognized by mAb 5E6 (Cat. No. 553277). A Ly-49I[BALB] allele has not been found. The Ly-49 family members are expressed independently, such that an individual NK or T cell may display more than one class of Ly-49 receptor homodimers. Binding of Ly-49I-expressing transfectants to cell lines bearing MHC class I antigens of the b,d, k and s haplotypes was not detected. In contrast, Ly-49I-expressing transfectants were found to weakly bind lymphoblasts of the b, d, k, q, s, and v haplotypes and to strongly bind H-2[r] lymphoblasts. Ly-49I-bearing NK cells may mediate allogeneic and hybrid resistance to transplantation of H-2[d] bone marrow.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49I
Applications Flow Cytometry
Classification Monoclonal
Clone YLI-90
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.