Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-49l Antibody (YLI-90), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128087
Description
Ly-49l Monoclonal specifically detects Ly-49l in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-49l | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells | |
Klra9 | |
CHO-K1 cells transfected with B6 allele of Ly49I gene | |
0.25 mg | |
Primary | |
B6 allele of the Ly49I receptor | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
YLI-90 | |
FITC | |
Killer cell lectin like receptor subfamily A member 9, Klra9, Ly49I receptor, LY49I1 | |
Mouse | |
Protein A or G purified | |
RUO | |
16640 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction