Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-51 Rat anti-Mouse, FITC, Clone: 6C3, BD

Catalog No. BDB562057 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562057 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562057 Supplier BD Biosciences Supplier No. 562057
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6C3 antibody reacts with an epitope of the 6C3/BP-1 (Ly-51) glycoprotein cell-surface differentiation antigen, which was originally identified on pre-B cell lymphomas (spontaneous and chemical- or retrovirus- transformed, in vitro and in vivo). 6C3/BP-1 is a homodimer cell-surface glycoprotein with 140-kDa subunits which has been identified to possess aminopeptidase A (APA) activity. The same antigen is expressed at high levels on bone marrow stromal cell lines which support in vitro B lymphopoieses, on thymic dendritic cells and cortical epithelial cells, and on a wide variety of mouse and rat tissues known to possess APA activity. Subsets of normal bone marrow pre-B and B lymphocytes express low levels of Ly-51, which is rapidly up-regulated on the pre-B cells in the presence of IL-7. A role for the 6C3/BP-1 molecule in the IL-7-driven proliferation of B cell precursors has been postulated. However, B-cell abnormalities were not detected in Ly-51-deficient mice. Mature B lymphocytes, thymocytes, peripheral T lymphocytes, erythroid cells, and myeloid cells (with the exception of thymic dendritic cells) do not express Ly-51. The 6C3 antibody can be used to identify cortical epithelium in frozen sections of thymuses from normal, SCID, and TCR-transgenic mice. It is possible that the low level of 6C3/BP-1 antigen detected, by flow cytometry, on some thymocytes may be passively adsorbed from adjacent epithelial cells during preparation of the cell suspensions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-51
Applications Flow Cytometry
Classification Monoclonal
Clone 6C3
Concentration 0.5mg/mL
Conjugate FITC
Description 6C3/BP-1 Antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen C57L mouse Pre-B lymphhoma cell line L1-2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.