Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6A/E Monoclonal Antibody (D7), PE

Catalog No. PIMA517891
Change view
Click to view available options
Quantity:
300 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517891 300 μg
PIMA517890 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517891 Supplier Invitrogen™ Supplier No. MA517891
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6A/E Monoclonal Antibody for Flow

Stem Cell Antigen-1 (Sca-1) is a member of the lymphocyte antigen 6 (Ly-6) family. It is expressed on multipotent hematopoietic stem cells (HSC) and is used as a marker of HSC in mice. Sca-1 is used in combination with lineage depletion antibodies to identify murine HSC. Sca-1 can be found in fetal liver, adult bone marrow, and adult peripheral blood and spleen. Sca-1 may be involved in the regulation of B and T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E
Applications Flow Cytometry
Classification Monoclonal
Clone D7
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Ly6a
Gene Accession No. P05533, Q64253
Gene Alias 9804; locus A/E; Locus AE; Ly6; Ly67; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly6A/E; Ly-6A/E; ly6a-e; Ly6e; Ly-6E; Ly-6E.1; lymphocyte antigen 6 complex; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6A-2/6E-1; Lymphocyte antigen 6E; RIG-E; Sca1; Sca-1; Sca-2; stem cell antigen 1; Stem cell antigen 2; TAP; T-cell-activating protein; thymic shared antigen 1; Tsa1; TSA-1
Gene Symbols Ly6a, Ly6e
Host Species Rat
Immunogen Mouse Ly-6A/E.
Purification Method Protein G
Quantity 300 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110454, 17069
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.