Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6A/E Rat anti-Mouse, Brilliant Violet 510, Clone: E13-161.7, BD Optibuild™

Catalog No. BDB744323 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744323 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744323 Supplier BD Biosciences Supplier No. 744323
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The E13-161.7 mAb recognizes Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Ly-6A/E is also known as stem cell antigen 1 (Sca-1/Sca1), or T-cell-activating protein (TAP). Ly-6A/E is a phosphatidylinositol-anchored protein of ∽18kDa that is expressed on multipotent hematopoietic stem cells (HSC) in mice with both Ly-6 haplotypes. Sca-1+ HSC are found in the adult bone marrow and fetal liver, but not in the early embryo yolk sac or intraembryonic hematopoietic sites, and can be mobilized to the peripheral blood and spleen in the adult. In mice expressing the Ly-6.2 haplotype, Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes, myeloid cells, and thymic and peripheral T lymphocytes, on the earliest intrathymic T-cell precursor population, and in several non-hematopoietic tissues. Strains with the Ly-6.1 haplotype have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of T cells from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb can block binding of FITC-conjugated D7 antibody (anti-Ly-6A/E) to mouse splenocytes, but purified mAb D7 is unable to block binding of FITC-conjugated E13-161.7 antibody.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E
Classification Monoclonal
Clone E13-161.7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SCA-1, Sca1, Stem cell antigen 1, TAP, T-cell-activating protein, Ly6
Host Species Rat
Immunogen BALB/c mouse-derived “pre-T” cell hybridoma
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.