Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6A/E Rat anti-Mouse, PE, Clone: D7, BD

Catalog No. BDB553108 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553108 0.2 mg
BDB562059 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553108 Supplier BD Biosciences Supplier No. 553108
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The D7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca-1 (Ly6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cells (HSC) in the bone mar

Host Species: Rat
Clone: D7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: IL-2-dependent mouse T-cell line CTL-L

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E
Applications Flow Cytometry
Classification Monoclonal
Clone D7
Concentration 0.2mg/mL
Conjugate PE
Description Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen IL-2-dependent mouse T-cell line CTL-L
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.