Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6A/E Rat anti-Mouse, PE-Cy7, Clone: D7, BD

Catalog No. BDB558162 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558162 0.1 mg
BDB561021 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB558162 Supplier BD Biosciences Supplier No. 558162
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The D7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca-1 (Ly6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cells (HSC) in the bone marrow of mice with both Ly-6 haplotypes. In mice expressing the Ly-6.2 haplotype (e.g., AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129), Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes and thymic and peripheral T lymphocytes. Strains with the Ly-6.1 haplotype (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of lymphocytes from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb (anti-Ly-6A/E, Mfr. No.553333) can block binding of FITC-conjugated D7 antibody (Mfr. No.557405) to mouse splenocytes, but purified mAb D7 is unable to block binding of FITC-conjugated E13-161.7 antibody (Mfr. No.553335). Anti-Ly-6A/E (Sca-1) mAb may be used in combination with the Mouse Lineage Panel (Mfr. No.559971) to identify HSC. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: D7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: IL-2-dependent mouse T-cell line CTL-L

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A/E
Applications Flow Cytometry
Classification Monoclonal
Clone D7
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Description Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen IL-2-dependent mouse T-cell line CTL-L
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.