Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102980 Supplier Invitrogen™ Supplier No. PA5102980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LY6A.

LY6A is included in a family of proteins that are highly homologous, low molecular weight glycophosphotidylinositol-linked proteins expressed on hematopoietic cells and lymphoid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ly6a
Gene Accession No. P05533
Gene Alias Ly6; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly-6A/E; Ly-6E.1; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6A-2/6E-1; Sca1; SCA-1; Stem cell antigen 1; TAP; T-cell-activating protein
Gene Symbols Ly6a
Host Species Rabbit
Immunogen A synthesized peptide derived from mouse LY6A(Accession P05533), corresponding to amino acid residues Q56-P76.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110454
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.