Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6A Recombinant Rabbit Monoclonal Antibody (JM53-11)
GREENER_CHOICE

Catalog No. PIMA541224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541224 Supplier Invitrogen™ Supplier No. MA541224
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Ly-6A Recombinant Monoclonal Antibody for IHC (P), Western Blot

LY6A is included in a family of proteins that are highly homologous, low molecular weight glycophosphotidylinositol-linked proteins expressed on hematopoietic cells and lymphoid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JM53-11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Ly6a
Gene Accession No. P05533
Gene Alias Ly6; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly-6A/E; Ly-6E.1; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6A-2/6E-1; Sca1; SCA-1; Stem cell antigen 1; TAP; T-cell-activating protein
Gene Symbols Ly6a
Host Species Rabbit
Immunogen Synthetic peptide within Mouse Ly-6A aa 41-90.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110454
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.