Learn More
Invitrogen™ Ly-6B.2 Monoclonal Antibody (7/4), PE
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA516542
Description
Reconstitute with 5 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Ly-6B.2 monoclonal antibody, clone 7/4 recognizes the Ly-6B.2 antigen.
Ly-6B.2 recognizes the Ly-6B.2 antigen. Ly-6B.2 is approximately 25-30 kDa, GPI-anchored, heavily glycosylated protein expressed on neutrophils, inflammatory monocytes and some activated macrophages. High levels of expression are seen in bone marrow, spleen, lung and lymph nodes. N-glycanase treatment of thioglycollate elicited peritoneal neutrophil lysates lowers the apparent molecular weight of Ly-6B.2 to approximately 15 kDa.
Specifications
Ly-6B.2 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
0 | |
Rat | |
Protein G | |
RUO | |
110453 | |
4°C, store in dark, DO NOT FREEZE! | |
Lyophilized |
Flow Cytometry | |
7/4 | |
PE | |
Ly6b | |
Ly6b | |
Cultured bone marrow cells. | |
500 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.