Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6B.2 Monoclonal Antibody (7/4)
GREENER_CHOICE

Catalog No. PIMA516539
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516539 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516539 Supplier Invitrogen™ Supplier No. MA516539
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Ly-6B.2 monoclonal antibody, clone 7/4 recognizes the Ly-6B.2 antigen.

Ly-6B.2 recognizes the Ly-6B.2 antigen. Ly-6B.2 is approximately 25-30 kDa, GPI-anchored, heavily glycosylated protein expressed on neutrophils, inflammatory monocytes and some activated macrophages. High levels of expression are seen in bone marrow, spleen, lung and lymph nodes. N-glycanase treatment of thioglycollate elicited peritoneal neutrophil lysates lowers the apparent molecular weight of Ly-6B.2 to approximately 15 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6B.2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7/4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ly6b
Gene Accession No. 0
Gene Symbols Ly6b
Host Species Rat
Immunogen Cultured bone marrow cells.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110453
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.