Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6C Antibody (ER-MP20), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065413APCCY7
Description
Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6C | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LY6C1 | |
Balb/c macrophage precursor cell hybrids | |
0.1 mL | |
17067.0 | |
Mouse | |
IgG2a |
Flow Cytometry | |
ER-MP20 | |
Flow Cytometry | |
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C | |
Rat | |
Protein G purified | |
Primary | |
Ly6C (ER-MP20) | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction