Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6C Antibody (ER-MP20), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065413UV
Description
Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Ly-6C | |
Monoclonal | |
DyLight 350 | |
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C | |
Rat | |
Protein G purified | |
RUO | |
17067 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
ER-MP20 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
Ly6c1 | |
Balb/c macrophage precursor cell hybrids | |
0.1 mL | |
Primary | |
Ly6C (ER-MP20) | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction