Learn More
Ly-6C Antibody (HK1.4), DyLight 755, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP128046IR
Catalog No. NBP128046IR
Only null left
Add to Cart
Description
Description
Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Ly-6C
Monoclonal
DyLight 755
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Rat
Protein A or G purified
RUO
17067
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
HK1.4
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
LY6C1
L3 cloned CTL cells
0.25 mL
Primary
Murine Ly-6C, Mr 18 kDa
Store at 4C in the dark.
IgG2c
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only