Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6C Rat anti-Mouse, BV480, Clone: HK1.4.rMAb (also known as HK1.4), BD Horizon™

Catalog No. BDB569438 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569438 0.1 mg
BDB569439 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569438 Supplier BD Biosciences Supplier No. 569438
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6C
Applications Flow Cytometry
Classification Monoclonal
Clone HK1.4.rMAb (also known as HK1.4)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly-6C; Ly-6C1; Ly6c; Ly6c1; Lymphocyte antigen Ly-6C
Host Species Rat
Immunogen L3 Clone CTL Cells
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.