Learn More
Ly-6C Rat anti-Mouse, FITC, Clone: AL-21, BD
Rat Monoclonal Antibody
$100.40 - $312.00
Specifications
Antigen | Ly-6C |
---|---|
Clone | AL-21 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561085
|
BD Biosciences
561085 |
25 μg |
Each for $100.40
|
|
|||||
BDB553104
|
BD Biosciences
553104 |
0.5 mg |
Each for $312.00
|
|
|||||
Description
The AL-21 monoclonal antibody specifically binds to a non-polymorphic determinant of Ly-6C, a 14-17 kDa GPI-linked cell-surface antigen found on some monocyte/macrophage populations, granulocytes, endothelial cells, plasma cells, and thymocyte, NK-cell, and T-subsets. Mice with the Ly-6.2 alloantigen (eg, AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129) have subsets of CD8+ and CD4+ Ly-6C+ T cells, while Ly-6.1 strains (eg, A, BALB/c, CBA, C3H/He, DBA/1, NZB) have only CD8+ Ly-6C+ T cells.Upregulation of Ly-6C expression on CD8+ T cells by interferons α and β and poly (I:C) has been described,and Ly-6C is a memory marker on CD8+ T cells.
Specifications
Ly-6C | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Not reported | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
AL-21 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgM κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.