Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6E Antibody, Novus Biologicals™
SDP

Catalog No. NBP152176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152176 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152176 Supplier Novus Biologicals Supplier No. NBP152176
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Ly-6E Polyclonal specifically detects Ly-6E in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen Ly-6E
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002337.1
Gene Alias lymphocyte antigen 6 complex, locus E, Retinoic acid-induced gene E protein, RIGE9804, RIG-Ely-6E, SCA2Ly-6E, SCA-2lymphocyte antigen 6E, Stem cell antigen 2, Thymic shared antigen 1, TSA1, TSA-1retinoic acid induced gene E
Gene Symbols LY6E
Host Species Goat
Immunogen Peptide with sequence SCQEKDHYCIT corresponding to internal region according to NP_002337.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4061
Test Specificity Reported variants represent identical protein: NP_002337.1, NP_001120685.1
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.