Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112981
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112981 Supplier Invitrogen™ Supplier No. PA5112981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ly-6E Polyclonal Antibody for Western Blot, ELISA

Sca1, also known as Ly6A/E, is a member of the Ly6 multigene family of type V glycophosphatidylinositol anchored cell surface proteins. It is expressed on multipotent hematopoietic stem cells in bone marrow of mice with both the Ly6.1 and Ly6.2 allotypes. In the periphery, Sca1 exhibits a pattern of expression which is based on differences between the two allotypes. Ly6.1 strains (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Sca1+ resting peripheral lymphocytes, whereas Ly6.2 strains (e.g., AKR, C57BL, C57BR, C57L, DBA/2, PL, SJL, SWR, 129) have relatively high numbers of Sca1+ lymphocytes. The expression of Sca1 is dramatically upregulated in all strains upon cellular activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6E
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LY6E
Gene Accession No. Q16553, Q64253
Gene Alias 9804; locus A/E; Locus AE; Ly67; Ly6A; Ly6e; ly-6E; lymphocyte antigen 6 complex, locus E; lymphocyte antigen 6E; retinoic acid induced gene E; retinoic acid-induced gene E protein; RIGE; RIG-E; SCA2; SCA-2; Stem cell antigen 2; thymic shared antigen 1; Tsa1; Tsa-1
Gene Symbols LY6E
Host Species Rabbit
Immunogen Recombinant Mouse Lymphocyte antigen 6E protein (21-102AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17069, 4061
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.