Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539231
Description
Ly-6A/E molecules are implicated in the regulation of T cell activation.
Specifications
| Ly-6E | |
| Polyclonal | |
| Unconjugated | |
| LY6E | |
| lymphocyte antigen 6 complex, locus A/E, Locus AE, Ly6A, Ly6E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4061 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16553 | |
| LY6E | |
| A synthetic peptide derived from the internal region of human LY6E | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction