Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RB6-8C5), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311978APC
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow CytometrySpecifications
Ly-6G/Ly-6C | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Flow Cytometry | |
RB6-8C5 | |
Flow Cytometry | |
GR, Gr-1, Ly-6, Ly6G | |
This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol | |
0.1 mL | |
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
546644 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction