Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RB6-8C5), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200441PE
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
LY6G | |
Normal mouse bone marrow. | |
0.1 mL | |
Primary | |
Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells. | |
Store at 4°C. Do not freeze. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay | |
RB6-8C5 | |
Flow Cytometry 1:10 - 1:1000 | |
GR, Gr-1, Ly-6, Ly6G | |
Rat | |
Protein A or G purified | |
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
546644.0 | |
Mouse | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction