Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RM0064-6D17), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211784APCCY7
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LY6G | |
Cell lysates of mouse neutrophils Ig | |
0.1 mL | |
Primary | |
This antibody detects mouse Ly-6G (Gr-1), the LY-6G/LY-6C complex. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
RM0064-6D17 | |
Flow Cytometry | |
GR, Gr-1, Ly-6, Ly6G | |
Rat | |
Protein A or G purified | |
Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
546644.0 | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction