Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RM0064-6D17) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$207.00 - $560.50
Specifications
| Antigen | Ly-6G/Ly-6C |
|---|---|
| Clone | RM0064-6D17 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, CyTOF, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Monoclonal |
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Ly-6G/Ly-6C | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| 546644 | |
| Cell lysates of mouse neutrophils Ig | |
| Primary | |
| This antibody detects mouse Ly-6G (Gr-1), the LY-6G/LY-6C complex. |
| RM0064-6D17 | |
| Immunohistochemistry, CyTOF, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| LY6G | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title