Learn More
Invitrogen™ Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Brilliant Ultra Violet™ 615, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | Ly-6G/Ly-6C |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RB6-8C5 |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Ultraviolet 615 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Ly6g |
| Gene Accession No. | P0CW02, P0CW03, P35461 |
| Gene Alias | AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.