Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE
Supplier: Thermo Scientific MA183934
Description

Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P0CW02, P0CW03, P35461 | |
Ly6c1, Ly6c2, Ly6g | |
Normal murine bone marrow cells. | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
RB6-8C5 | |
PE | |
Ly6c2 | |
AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
Rat | |
Protein G | |
RUO | |
100041546, 17067, 546644 | |
4°C, store in dark, DO NOT FREEZE! | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction