Learn More
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$165.00 - $252.00
Specifications
| Antigen | Ly-6G/Ly-6C |
|---|---|
| Clone | RB6-8C5 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9684
|
Invitrogen™
25593181 |
50 μg |
Each for $165.00
|
|
|||||
50-112-9597
![]()
|
Invitrogen™
25593182 |
100 μg |
Each for $252.00
|
|
|||||
Description
Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets. Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation.
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.Specifications
| Ly-6G/Ly-6C | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
| Ly6c1, Ly6c2, Ly6g | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| RB6-8C5 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| Rat | |
| Mouse | |
| P0CW02, P0CW03, P35461 | |
| 100041546, 17067, 546644 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.