Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G/Ly-6C, Rat Anti-Mouse, RB613, Clone: RB6-8C5, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 759334
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
Ly-6G/Ly-6C | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg; | |
Primary | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
RB6-8C5 | |
RB613 | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
Not Reported | |
RUO | |
17067, 546644 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction