Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ly-6G/Ly-6C Recombinant Rat Monoclonal Antibody (RB6-8C5)
Rat Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548136
Description
Specificity: This antibody is specific for murine Ly6G, a a 21-25 kDa glycosylphosphatidylinositol (GPI)-linked protein expressed by the myeloid-derived cells in a tightly developmentally-regulated manner in the bone marrow.
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications
Ly-6G/Ly-6C | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P0CW02, P0CW03, P35461 | |
Ly6c1, Ly6c2, Ly6g | |
This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol as described by Martiniello et al (1994). | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry, Functional Assay, Immunohistochemistry, Western Blot | |
RB6-8C5 | |
Unconjugated | |
Ly6c2 | |
AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
Rat | |
Protein A | |
RUO | |
100041546, 17067, 546644 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction