Learn More
Ly-6G Rat anti-Mouse, FITC, Clone: 1A8, BD
Rat Monoclonal Antibody
$78.00 - $307.50
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561105
|
BD Biosciences
561105 |
25 μg |
Each for $78.00
|
|
|||||
BDB551460
|
BD Biosciences
551460 |
0.5 mg |
Each for $307.50
|
|
|||||
Description
The 1A8 monoclonal antibody specifically binds to Ly-6G, a 21-25-kDa glycosyolphosphatidylinositol-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The RB6-8C5 monoclonal antibody recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Ly-6G | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Ly-6G-transfected EL4J cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
1A8 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.