Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G Rat anti-Mouse, RY586, Clone: 1A8, BD Horizon™
Rat Monoclonal Antibody
$158.40 - $394.00
Specifications
| Antigen | Ly-6G |
|---|---|
| Clone | 1A8 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Ly-6G Monoclonal antibody specifically detects Ly-6G in Mouse samples. It is validated for Flow cytometry (Routinely Tested)Specifications
| Ly-6G | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Ly-6G-transfected EL4J Cell Line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
| 1A8 | |
| Flow Cytometry | |
| RY586 | |
| Rat | |
| Mouse | |
| Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1 | |
| IgG2a κ | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title