Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6G6F Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Ly-6G6F |
---|---|
Dilution | Western Blot 1:100-1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Ly-6G6F Polyclonal specifically detects Ly-6G6F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ly-6G6F | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
259215 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LLGNYSLWLEGSKEEDAGRYWCAVLGQHHNYQNWRVYDVLVLKGSQLSARAADGSPCNVLLCSVVPSRRMDSVTWQEGKGPVRGRVQSFWGSEAALLLVCPGEGLSEPRSRRPRIIRCLMTHNKGVSFSLAASIDASPAL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:100-1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Unconjugated | |
RUO | |
Human | |
C6orf21, G6F, locus G6D, LY6G6D, lymphocyte antigen 6 complex locus protein G6f, lymphocyte antigen 6 complex, locus G6F, NG32 | |
LY6G6F | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title