Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ly108/Slamf6, Rat, Unlabeled, Clone: 3E11,

Catalog No. MABF919MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF919MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF919MI Supplier MilliporeSigma™ Supplier No. MABF919
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SLAM family member 6 (UniProt Q9ET39; also known as CD352, NK-T-B-antigen, NTB-A, Lymphocyte antigen 108) is encoded by the Slamf6 (also known as Ly108) gene (Gene ID 30925) in murine species. Ly108/Slamf6 belongs to the 7-member (SLAM/CD150, CD48/SLAMF2, Ly-9/CD229, 2B4/CD244, CD84/SLAMF5, Ly108/NTB-A/CD352, CRACC) signaling lymphocytic activation molecule family (SLAMF) of receptors broadly expressed in hematopoietic cells. 2B4 and CD48 interact with eath other, while rest of the family members act as self-ligands, where two identical molecules on the surface of two different cells can interact as receptor and ligand to each other. SLAMF members carry one or more copies of immunoreceptor tyrosine-based switch motif or ITSM (TxYxxV/I) in their cytoplasmic tails. Upon phosphorylation on the tyrosine residue, ITSM mediates the recruitment of SH2 domain-containing signaling molecules such as SLAM-associated protein (SAP), including SAP in T-, NK, and NK T-cells, EAT-2A and EAT-2B (murine) in NK cells and APCs. Ly108/Slamf6 is synthesized with a signal peptide sequence (mouse a.a. 1-30), the removal of which yields the mature Ly108 with an extracellular region (a.a. 31-239) that contains a V-type and a C2-type Ig-like domain (a.a. 36-130 and 147-210), followed by a single transmembrane domain (a.a. 240-262) and a cytoplasmic tail (a.a. 263-351). Ly108/Slamf6 cytoplasmic domain contains two copies of ITSM (mouse a.a. 293-TVYAQV-298 and 317-TIYSIV-322). Alternative splicings produce three Ly108 isoforms, Ly108-1 (Ly108s; UniProt Q9ET39-2), Ly108-2 (Ly108l; UniProt Q9ET39-1) and Ly108-3 (Not reported by UniProt), that differ in their cytoplasmic C-terminal end sequence, but are identical in the rest of their sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly108/Slamf6
Applications Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3E11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2a in buffer containing PBS without preservatives.
Gene Slamf6, Ly108
Gene Accession No. Q9ET39
Host Species Rat
Immunogen Murine Ly108-human IgG1 Fc fusion protein.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_109635.1
Test Specificity Clone 3E11 specifically detected exogenously expressed murine Ly108-1 (Ly108s; UniProt Q9ET39-2) and Ly108-2 (Ly108l; UniProt Q9ET39-1), but not other SLAM family members (2B4, CD84, CRACC, Ly-9, and SLAM) on the surface of respective mouse BI-141 T cell transfectants. Murine Ly108 spliced isoforms Ly108-1 (Ly108s; UniProt Q9ET39-2), Ly108-2 (Ly108l; UniProt Q9ET39-1) and Ly108-3 (Not reported by UniProt) differ in their cytoplasmic C-terminal end sequence, but are identical in the rest of their sequence, including the extracellular region targeted by clone 3E11 (Zhong, M.C., and Veillette, A. (2008). J. Biol. Chem. 283(28):19255 -19264).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.