Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly49A Mouse anti-Mouse, Unconjugated, Clone: A1, MilliporeSigma™

Catalog No. MABF209025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2090-25 25 μg
MABF2090-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2090-25 Supplier MilliporeSigma Supplier No. MABF209025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Ly49A, clone A1, Cat. No. MABF2090 is tested for use in Flow Cytometry, Immunocytochemistry and Immunoprecipitation, for the detection of Lymphocyte antigen 49a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly49A
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone A1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias T-cell surface glycoprotein YE1/48;Lymphocyte antigen 49a;Ly-49a;T lymphocyte antigen A1
Gene Symbols Klra1;LY-49;Ly-49a;LY49A
Host Species Mouse
Immunogen EL-4 T lymphoma cells.
Molecular Weight of Antigen 30.50 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone A1 specifically detects Lymphocyte antigen 49a (LY49A) in murine species.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.