Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly6al Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579616
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579616 Supplier Invitrogen™ Supplier No. PA579616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Cardiac Muscle Tissue, Rat Liver Tissue.

Ly6al, lymphocyte antigen 6 complex, locus A-like, is also known as Ly6a, Ly6a_predicted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly6al
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Ly6al
Gene Accession No. 0
Gene Alias Ly6a; Ly6-A antigen; Ly6al; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6 complex, locus A-like
Gene Symbols Ly6al
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of rat Ly6al (91-108aa, TVVQVNTSCCTRDLCNAA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362935
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.