Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564167 Supplier Invitrogen™ Supplier No. PA564167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESCTPSYTLQ GQVSSGTSST QCCQEDLCNE KLHNAAP Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 59%.

May act as a specification marker at earliest stage specification of lymphocytes between B- and T-cell development. Marks the earliest stage of B-cell specification. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ly6d
Gene Accession No. Q14210
Gene Alias E48; E48 antigen; Ly61; Ly-61; LY6D; Ly-6D; lymphocyte antigen 6 complex, locus D; lymphocyte antigen 6D; Thb; thymocyte B-cell antigen
Gene Symbols Ly6d
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LY6D. Recombinant protein control fragment (Product #RP-103907).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.