Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6g, Rat anti-Mouse, Clone: Nimp-R14, Abnova™

Catalog No. 89998884 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-884 Supplier Abnova Corporation Supplier No. MAB16109
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifications

Antigen Ly6g
Applications Blocking Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Nimp-R14
Conjugate Unconjugated
Dilution Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Immunohistochemistry (Frozen sections) (1:200) Immunocytochemistry (1:200) Blocking (250 ug/mouse) The optimal working dilution should be determined by the end user.
Formulation In PBS (10% glycerol, 0.02% sodium azide).
Gene lymphocyte antigen 6 complex, locus G
Gene Alias Gr-1/Gr1/Ly-6G
Gene Symbols Ly6g
Host Species Rat
Immunogen Purified mouse BALB/c neutrophils of surface protein Ly6g.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 546644
Target Species Mouse
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.