Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY6G Rat anti-Mouse, PE-Cy7, Clone: RB6-8C5, Abnova™

Catalog No. 89934495 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-495 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-495 Supplier Abnova Corporation Supplier No. MAB12236
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Ly6g.

Specifications

Antigen LY6G
Applications Flow Cytometry
Classification Monoclonal
Clone RB6-8C5
Conjugate PE-Cyanine7
Description Rat monoclonal antibody raised against Ly6g.
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer).
Gene Alias Gr-1/Gr1/Ly-6G
Gene Symbols Ly6g
Host Species Rat
Immunogen Native purified from normal murine bone marrow cells.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 546644
Target Species Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.