Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6G5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561705 Supplier Invitrogen™ Supplier No. PA561705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CCQYDYCNSW SSPQLQSSLP EPHDRPLALP LSDSQIQWFY QALNLSLPLP NFHAGTEPDG LDPMVTLSLN LGLSFAELRR MYLFLNSSGL LVLPQ Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 61%.

LY6G5B belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6G5B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LY6G5B
Gene Accession No. Q8NDX9
Gene Alias C6orf19; G5B; LY6G5B; Lymphocyte antigen 6 complex locus protein G5b; lymphocyte antigen 6 complex, locus G5B; lymphocyte antigen 6 family member G5B; lymphocyte antigen-6 G5B splicing isoform 288; lymphocyte antigen-6 G5B splicing isoform 452; lymphocyte antigen-6 G5B splicing isoform 690; lymphocyte antigen-6 G5B splicing isoform 837
Gene Symbols LY6G5B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LY6G5B. Recombinant protein control fragment (Product #RP-100842).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58496
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.