Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6G5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558495 Supplier Invitrogen™ Supplier No. PA558495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADLPSCWGAG PCYTGHKVGA LRRDTVICCC RHGDYSTPCL FTPGKPSRNP SPWKRTLWT Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.

LY6G5C belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6G5C
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ly6g5c
Gene Accession No. Q5SRR4
Gene Alias C6orf20; Canine epididymal gene product 8; CE8; CE8 protein, variant 2; epididymal secretory protein 8; G5C; Ly6g5c; LY6G5CA; LY6G5CB; Lymphocyte antigen 6 complex locus protein G5c; lymphocyte antigen 6 complex, locus G5C; lymphocyte antigen 6 family member G5C; lymphocyte antigen-6 G5C; NG33; re8
Gene Symbols Ly6g5c
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LY6G5C. Recombinant protein control fragment (Product #RP-96588).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.