Learn More
Invitrogen™ LY6G6C Polyclonal Antibody

Description
LY6G6C Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | LY6G6C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LY6G6C |
| Gene Accession No. | O95867 |
| Gene Alias | C6orf24; G6C; LY6G6C; Lymphocyte antigen 6 complex locus protein G6c; lymphocyte antigen 6 complex, locus G6C; lymphocyte antigen-6 G6C; NG24; UNQ1947/PRO4430 |
| Gene Symbols | LY6G6C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.